Learn More
GCC2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GCC2.
Brand: Abnova PAB22917.100uL
Description
The protein encoded by this gene is a peripheral membrane protein localized to the trans-Golgi network. It is sensitive to brefeldin A. This encoded protein contains a GRIP domain which is thought to be used in targeting. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: QNISEANSQHYQKNINSLQEELLQLKAIHQEEVKELMCQIEASAKEHEAEINKLNELKENLVKQCEASEKNIQKKYECELENLRKATSNANQDNQICSILSpecifications
| GCC2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GCC2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q8IWJ2 | |
| GCC2 | |
| Recombinant protein corresponding to amino acids of human GCC2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| GCC2 | |
| GCC185/KIAA0336 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9648 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only