Learn More
GBP1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GBP1.
Brand: Abnova PAB25097.100uL
Description
Guanylate binding protein expression is induced by interferon. Guanylate binding proteins are characterized by their ability to specifically bind guanine nucleotides (GMP, GDP, and GTP) and are distinguished from the GTP-binding proteins by the presence of 2 binding motifs rather than 3. [provided by RefSeq
Specifications
| GBP1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| GBP1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 2633 | |
| Human | |
| Liquid |
| Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of GBP1. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| GBP1 | |
| A synthetic peptide corresponding to GBP1. | |
| 100 μL | |
| Primary | |
| GBP1 polyclonal antibody detects endogenous levels of GBP1 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only