missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA6, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GATA6.
Brand: Abnova PAB4926.400uL
This item is not returnable.
View return policy
Specifications
| GATA6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of GATA6. | |
| In PBS (0.09% sodium azide) | |
| NP_005248;Q92908 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 2627 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Dot Blot, ELISA | |
| Unconjugated | |
| ELISA (1:1000) Dot Blot (1:500) The optimal working dilution should be determined by the end user. | |
| GATA6 | |
| GATA6 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y271 of human GATA6. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction