Learn More
GAPDH Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant GAPDH.
Brand: Abnova PAB17013.100ug
Description
The product of this gene catalyzes an important energy-yielding step in carbohydrate metabolism, the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD). The enzyme exists as a tetramer of identical chains. Many pseudogenes similar to this locus are present in the human genome. [provided by RefSeq
Specifications
| GAPDH | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant GAPDH. | |
| In 20mM PBS, 0.15M NaCl, pH 7.2 (0.01% sodium azide) | |
| G3PD/GAPD/MGC88685 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000-1:4000) Immunofluorescence (1:100-1:400) Immunohistochemistry (1:100-1:400) The optimal working dilution should be determined by the end user. | |
| GAPDH | |
| GAPDH | |
| Recombinant protein corresponding to human GAPDH. | |
| RUO | |
| 2597 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only