Learn More
GALNT8, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GALNT8.
Brand: Abnova PAB20609.100uL
Description
This gene encodes a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes. GalNAc-Ts initiate mucin-type O-linked glycosylation in the Golgi apparatus by catalyzing the transfer of GalNAc to serine and threonine residues on target proteins. They are characterized by an N-terminal transmembrane domain, a stem region, a lumenal catalytic domain containing a GT1 motif and Gal/GalNAc transferase motif, and a C-terminal ricin/lectin-like domain. GalNAc-Ts have different, but overlapping, substrate specificities and patterns of expression. [provided by RefSeq
Sequence: NLLDENVCLDQGPFPGNTPIMYYCHEFSSQNVYYHLTGELYVGQLIAEASASDRCLTDPGKAEKPTLEPCSKAAKNRLHIYWDFKPGGAVINRDTKRCLEMKKDLLGSHVLVLQTCSTQVWEIQHTVRSpecifications
| GALNT8 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GALNT8. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9NY28 | |
| GALNT8 | |
| Recombinant protein corresponding to amino acids of human GALNT8. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| GALNT8 | |
| GALNAC-T8 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 26290 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only