Learn More
GAL Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GAL.
Brand: Abnova PAB5055.150ug
Description
Galanin is small neuropeptide that functions as a cellular messenger within the central and peripheral nervous systems, modulating diverse physiologic functions (Mechenthaler, 2008 [PubMed 18500643]).[supplied by OMIM
Sequence: GWTLNSAGYLLGPHAVGNHRSFSDKNGLTSSpecifications
| GAL | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GAL. | |
| In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal) | |
| GALN/GLNN/GMAP/MGC40167 | |
| Rabbit | |
| 150 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunoprecipitation | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| GAL | |
| GAL | |
| A synthetic peptide (conjugated with KLH) corresponding to human GAL. | |
| RUO | |
| 51083 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only