missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gabrb3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a recombinant Gabrb3.
Brand: Abnova PAB9687.100uL
This item is not returnable.
View return policy
Specifications
| Gabrb3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a recombinant Gabrb3. | |
| In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
| P63079 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 24922 | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Gabrb3 | |
| Gabrb3 | |
| Recombinant protein corresponding to rat Gabrb3. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction