Learn More
GAB1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GAB1.
Brand: Abnova PAB26803.100ug
Description
The protein encoded by this gene is a member of the IRS1-like multisubstrate docking protein family. It is an important mediator of branching tubulogenesis and plays a central role in cellular growth response, transformation and apoptosis. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: V-E-Yp-L-DSpecifications
| GAB1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| GAB1 | |
| GAB1 | |
| A synthetic peptide corresponding to residues surrounding Y627 of human GAB1. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of GAB1. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| Q13480 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2549 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only