Learn More
FZD3 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FZD3.
Brand: Abnova PAB26956.100uL
Description
This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. The function of this protein is unknown, although it may play a role in mammalian hair follicle development. [provided by RefSeq
Specifications
| FZD3 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| FZD3 | |
| Fz-3/hFz3 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 7976 | |
| Human, Mouse | |
| Liquid |
| Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of FZD3. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| Q9NPG1 | |
| FZD3 | |
| A synthetic peptide corresponding to human FZD3. | |
| 100 μL | |
| Primary | |
| FZD3 polyclonal antibody detects endogenous levels of FZD3 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only