Learn More
FYN Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FYN.
Brand: Abnova PAB27162.100uL
Description
This gene is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist. [provided by RefSeq
Specifications
| FYN | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| FYN | |
| MGC45350/SLK/SYN | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2534 | |
| Human, Mouse | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of FYN. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| P06241 | |
| FYN | |
| A synthetic peptide corresponding to human FYN. | |
| 100 μL | |
| Primary | |
| FYN polyclonal antibody detects endogenous levels of FYN protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only