Learn More
FXYD6, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FXYD6.
Brand: Abnova PAB24172.100uL
Description
This reference sequence was derived from multiple replicate ESTs and validated by human genomic sequence. This gene encodes a member of a family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 invariant and 6 highly conserved amino acids. The approved human gene nomenclature for the family is FXYD-domain containing ion transport regulator. FXYD2, also known as the gamma subunit of the Na,K-ATPase, regulates the properties of that enzyme. FXYD1 (phospholemman), FXYD2 (gamma), FXYD3 (MAT-8), FXYD4 (CHIF), and FXYD5 (RIC) have been shown to induce channel activity in experimental expression systems. Transmembrane topology has been established for two family members (FXYD1 and FXYD2), with the N-terminus extracellular and the C-terminus on the cytoplasmic side of the membrane. This gene product, FXYD6, is novel and has not been characterized as a protein. Multiple alternatively spliced transcript variants that encode the same protein isoform have been described. RefSeq curation by Kathleen J. Sweadner, Ph.D.
Sequence: SRRCKCSFNQKPRAPGDEEAQVENLITANATEPSpecifications
| FXYD6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FXYD6. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9H0Q3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 53826 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| FXYD6 | |
| FXYD6 | |
| Recombinant protein corresponding to amino acids of human FXYD6. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only