Learn More
FXYD5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FXYD5.
Brand: Abnova PAB20515.100uL
Description
This gene encodes a member of a family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 invariant and 6 highly conserved amino acids. The approved human gene nomenclature for the family is FXYD-domain containing ion transport regulator. Mouse FXYD5 has been termed RIC (Related to Ion Channel). FXYD2, also known as the gamma subunit of the Na,K-ATPase, regulates the properties of that enzyme. FXYD1 (phospholemman), FXYD2 (gamma), FXYD3 (MAT-8), FXYD4 (CHIF), and FXYD5 (RIC) have been shown to induce channel activity in experimental expression systems. Transmembrane topology has been established for two family members (FXYD1 and FXYD2), with the N-terminus extracellular and the C-terminus on the cytoplasmic side of the membrane. This gene product, FXYD5, is a glycoprotein that functions in the up-regulation of chemokine production, and it is involved in the reduction of cell adhesion via its ability to down-regulate E-cadherin. It also promotes metastasis, and has been linked to a variety of cancers. Alternative splicing results in multiple transcript variants. [RefSeq curation by Kathleen J. Sweadner, Ph.D.
Sequence: DTTSSSSADSTIMDIQVPTRAPDAVYTELQPTSPTPTWPADETPQPQTQTQQLEGTDGPLVTDPETHKSTKAAHPTDDTTTLSERPSPSTDVQTDPQTLKPSGFHEDDPFFYDEHTLRSpecifications
| FXYD5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FXYD5. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q96DB9 | |
| FXYD5 | |
| Recombinant protein corresponding to amino acids of human FXYD5. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| FXYD5 | |
| HSPC113/IWU-1/IWU1/KCT1/OIT2/PRO6241/RIC/dysad | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 53827 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only