Learn More
FRRS1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FRRS1.
Brand: Abnova PAB20970.100uL
Description
Members of the cytochrome b561 (CYB561; MIM 600019) family, including FRRS1, reduce ferric to ferrous iron before its transport from the endosome to the cytoplasm (Vargas et al., 2003 [PubMed 14499595]).[supplied by OMIM
Sequence: SHLTKPFSASDCGNKKFCIRSPLNCDPEKEASCVFLSFTRDDQSVMVEMSGPSKGYLSFALSHDQWMGDDDAYLCIHEDQTVYIQPSHLTGRSHPVMDSRDTLEDMAWRLADGVMQCSFRRNITLPGVKNRFDLNTSYYIFLADGAANDGSpecifications
| FRRS1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FRRS1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q6ZNA5 | |
| FRRS1 | |
| Recombinant protein corresponding to amino acids of human FRRS1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| FRRS1 | |
| SDFR2/SDR2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 391059 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only