Learn More
FRAT1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FRAT1.
Brand: Abnova PAB3083.400uL
Description
The protein encoded by this gene belongs to the GSK-3-binding protein family. The protein inhibits GSK-3-mediated phosphorylation of beta-catenin and positively regulates the Wnt signaling pathway. It may function in tumor progression and in lymphomagenesis. [provided by RefSeq
Specifications
| FRAT1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of FRAT1. | |
| In PBS (0.09% sodium azide) | |
| NP_005470;Q92837 | |
| FRAT1 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human FRAT1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| FRAT1 | |
| FLJ97193 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10023 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only