Learn More
FOXO3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FOXO3.
Brand: Abnova PAB5364.100ug
Description
This gene belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain. This gene likely functions as a trigger for apoptosis through expression of genes necessary for cell death. Translocation of this gene with the MLL gene is associated with secondary acute leukemia. Alternatively spliced transcript variants encoding the same protein have been observed. [provided by RefSeq
Specifications
| FOXO3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of FOXO3. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| AF6q21/DKFZp781A0677/FKHRL1/FKHRL1P2/FOXO2/FOXO3A/MGC12739/MGC31925 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| FOXO3 | |
| FOXO3 | |
| A synthetic peptide corresponding to residues surrounding S253 of human FOXO3. | |
| RUO | |
| 2309 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only