missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXN2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FOXN2.
Brand: Abnova PAB20217.100uL
This item is not returnable.
View return policy
Description
This gene encodes a forkhead domain binding protein and may function in the transcriptional regulation of the human T-cell leukemia virus long terminal repeat. [provided by RefSeq
Sequence: LSLNKCFQKVERSHGKVNGKGSLWCVDPEYKPNLIQALKKQPFSSASSQNGSLSPHYLSSVIKQNQVRNLKESDIDAAAAMMLLNTSIEQGILECEKPLPLKTALQKKRSYGNAFHHPSAVRLQESDSLATSIDPKEDHNYSASpecifications
| FOXN2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FOXN2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P32314 | |
| FOXN2 | |
| Recombinant protein corresponding to amino acids of human FOXN2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| FOXN2 | |
| HTLF | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3344 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction