Learn More
FOXH1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FOXH1.
Brand: Abnova PAB25674.100ug
Description
FOXH1 encodes a human homolog of Xenopus forkhead activin signal transducer-1. FOXH1 protein binds SMAD2 and activates an activin response element via binding the DNA motif TGT(G/T)(T/G)ATT. [provided by RefSeq
Specifications
| FOXH1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| FOXH1 | |
| FAST-1/FAST1 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 8928 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of FOXH1. | |
| In PBS (0.05% sodium azide) | |
| NP_003914 | |
| FOXH1 | |
| A synthetic peptide corresponding to 19 amino acids at N-terminus of human FOXH1. | |
| 100 μg | |
| Primary | |
| FOXH1 antibody is predicted to not cross-react with any other members of the forkhead box family. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only