missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXD4/FOXD4L2/FOXD4L3/FOXD4L4/FOXD4L5/FOXD4L6, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FOXD4/FOXD4L2/FOXD4L3/FOXD4L4/FOXD4L5/FOXD4L6.
Brand: Abnova PAB17720.100ug
This item is not returnable.
View return policy
Specifications
| FOXD4/FOXD4L2/FOXD4L3/FOXD4L4/FOXD4L5/FOXD4L6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of FOXD4/FOXD4L2/FOXD4L3/FOXD4L4/FOXD4L5/FOXD4L6. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| FKHL9/FOXD4A/FREAC5/MGC105106 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total FOXD4/FOXD4L2/FOXD4L3/FOXD4L4/FOXD4L5/FOXD4L6 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| Q3SYB3/Q12950/Q8WXT5/Q5VV16/Q6VB85/Q6VB84 | |
| FOXD4 | |
| A synthetic peptide corresponding to internal of human FOXD4/FOXD4L2/FOXD4L3/FOXD4L4/FOXD4L5/FOXD4L6. | |
| RUO | |
| 2298/100036519/286380/349334/653427/653404 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction