Learn More
FOSB Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FOSB.
Brand: Abnova PAB19547.100ug
Description
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| FOSB | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of FOSB. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| AP-1/DKFZp686C0818/G0S3/GOS3/GOSB/MGC42291 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 2354 | |
| Human | |
| Lyophilized |
| Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| FOSB | |
| FOSB | |
| A synthetic peptide corresponding to amino acids at N-terminus of human FOSB. | |
| 100 μg | |
| Primary | |
| The synthetic peptide identical to the related mouse and rat sequences. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only