Learn More
FOS Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a synthetic peptide of FOS.
Brand: Abnova PAB0828.200ug
Description
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. In some cases, expression of the FOS gene has also been associated with apoptotic cell death. [provided by RefSeq
Specifications
| FOS | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a synthetic peptide of FOS. | |
| In PBS (0.2% gelatin, 0.09% sodium azide) | |
| AP-1/C-FOS | |
| Rabbit | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:200) The optimal working dilution should be determined by the end user. | |
| FOS | |
| FOS | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human FOS. | |
| RUO | |
| 2353 | |
| Store at 4°C. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only