missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FN1 Rabbit anti-Chicken, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native FN1.
Brand: Abnova PAB13518.500uL
This item is not returnable.
View return policy
Specifications
| FN1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native FN1. | |
| Lyophilized from purified antibody | |
| FN1 | |
| Native purified FN1 from chicken plasma. | |
| RUO | |
| 396133 | |
| Chicken | |
| Lyophilized |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:600) Immunofluorescence (1:40) The optimal working dilution should be determined by the end user. | |
| FN1 | |
| Rabbit | |
| 500 μL | |
| Primary | |
| This antibody reacts with chicken fibronectin 100%, chicken collagen type I, III and V <0.1% (RIA at 1:10000 dilution). | |
| Store at 4°C on dry atmosphere. After reconstitution with 0.5mL distilled water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction