Learn More
FMNL1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FMNL1.
Brand: Abnova PAB25671.100ug
Description
This gene encodes a formin-related protein. Formin-related proteins have been implicated in morphogenesis, cytokinesis, and cell polarity. An alternative splice variant has been described but its full length sequence has not been determined. [provided by RefSeq
Specifications
| FMNL1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| FMNL1 | |
| C17orf1/C17orf1B/FHOD4/FMNL/KW-13/MGC133052/MGC1894/MGC21878 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 752 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of FMNL1. | |
| In PBS (0.05% sodium azide) | |
| NP_005883 | |
| FMNL1 | |
| A synthetic peptide corresponding to 16 amino acids at C-terminus of human FMNL1. | |
| 100 μg | |
| Primary | |
| FRL1 antibody is human, mouse and rat reactive. Three alternatively spliced transcript variants have been observed. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only