Learn More
FLAD1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FLAD1.
Brand: Abnova PAB22164.100uL
Description
This gene encodes the enzyme that catalyzes adenylation of flavin mononucleotide (FMN) to form flavin adenine dinucleotide (FAD) coenzyme. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq
Sequence: TNTFFLCRTLRSLGVQVCRVSVVPDEVATIAAEVTSFSNRFTHVLTAGGIGPTHDDVTFEAVAQAFGDELKPHPKLEAATKASpecifications
| FLAD1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FLAD1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q8NFF5 | |
| FLAD1 | |
| Recombinant protein corresponding to amino acids of human FLAD1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| FLAD1 | |
| FAD1/FADS/MGC31803/MGC40255/PP591/RP11-307C12.7 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 80308 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only