missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Fish Immunoglobulin Secondary Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-68352

Product Code. 18051106

  • £474.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunoglobulin Polyclonal specifically detects Immunoglobulin in Fish samples. It is validated for Western Blot, ELISA, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Immunoglobulin
Polyclonal
Western Blot, ELISA 1:5000 - 1:10000, Immunofluorescence
Rabbit
Whole Antiserum
Secondary
Fish
Antisera
Western Blot, ELISA, Immunofluorescence
Unconjugated
Serum with 0.09% Sodium Azide
Rainbow trout and Atlantic salmon immunoglobulins.
50 μL
Rabbit anti Salmonid Ig antibody recognizes salmonid immunoglobulin, binding to Ig of the Salmo, Oncorhynchus and Salvelinus genera
Store at 4°C.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.