Learn More
FIGF Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FIGF.
Brand: Abnova PAB17217.100ug
Description
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family and is active in angiogenesis, lymphangiogenesis, and endothelial cell growth. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-2 and VEGFR-3 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor C. [provided by RefSeq
Specifications
| FIGF | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of FIGF. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| VEGF-D/VEGFD | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 2277 | |
| Human | |
| Lyophilized |
| Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| FIGF | |
| FIGF | |
| A synthetic peptide corresponding to amino acids at internal region of human FIGF. | |
| 100 μg | |
| Primary | |
| The synthetic peptide of this antibody is identical to the related rat sequence. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only