missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen fragment E, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Fibrinogen fragment E.
Brand: Abnova PAB14914.1mL
This item is not returnable.
View return policy
Specifications
| Fibrinogen fragment E | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Fibrinogen fragment E. | |
| In antiserum | |
| Native purified plasmin digestion product E of human fibrinogen. | |
| RUO | |
| This antibody reacts with intact fibrinogen and fibrinogenin from human plasma. No cross-reactivity with human fibrinogen fragment D (when assessed with double diffusion/electrophoresis). | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Immunodiffusion (1:32) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction