Learn More
FGG, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against FGG.
Brand: Abnova PAB14903.1mL
Description
The protein encoded by this gene is the gamma component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia and thrombophilia. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq
Specifications
| FGG | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against FGG. | |
| In serum | |
| FGG | |
| Human FGG. | |
| RUO | |
| 2266 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunocytochemistry, Immunodiffusion, Immunomicroscopy, Radioimmune Assays (RIA) | |
| Unconjugated | |
| ELISA (1:10000) Immunodiffusion (1:32) The optimal working dilution should be determined by the end user. | |
| FGG | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only