Learn More
FER Rabbit anti-Dog, Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FER.
Brand: Abnova PAB15526.100uL
Description
Fer protein is a member of the FPS/FES family of nontransmembrane receptor tyrosine kinases. It regulates cell-cell adhesion and mediates signaling from the cell surface to the cytoskeleton via growth factor receptors. [provided by RefSeq
Specifications
| FER | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of FER. | |
| In PBS (30% glycerol, 0.09% sodium azide) | |
| P16591 | |
| FER | |
| A synthetic peptide corresponding to amino acids 400-450 of human FER. | |
| RUO | |
| 2241 | |
| Canine, Human, Mouse | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| FER | |
| TYK3 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Immunogen has 100% homology with mouse and canine. This antibody is useful in Western blot, where a band is seen ∼94 KDa. | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only