missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Feline IgG Fc Secondary Antibody, Rhodamine, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-73465
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Feline samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA.
Specifications
| IgG Fc | |
| Polyclonal | |
| Rhodamine | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| No reaction was observed against Cat IgG F(ab'). | |
| Feline | |
| IgG |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Cat IgG F(c) fragment | |
| 2 mg | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction