missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Feline IgG Fc Secondary Antibody, DyLight 350, Novus Biologicals™

Product Code. 30228403 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30228403 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30228403 Supplier Novus Biologicals Supplier No. NBP172771UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IgG Fc Polyclonal antibody specifically detects IgG Fc in Feline samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG Fc
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Host Species Rabbit
Immunogen Cat IgG F(c) fragment
Purification Method Multi-step
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Feline
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.