missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FDCSP, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FDCSP.
Brand: Abnova PAB20711.100uL
This item is not returnable.
View return policy
Description
FDCSP is expressed by follicular dendritic cells (FDCs) and activated leukocytes during ongoing immune responses.[supplied by OMIM
Sequence: KRSISDSDELASGFFVFPYPYPFRPLPPIPFPRFPWFRRNFPIPIPESAPTTPLSpecifications
| FDCSP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FDCSP. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q8NFU4 | |
| FDCSP | |
| Recombinant protein corresponding to amino acids of human FDCSP. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| FDCSP | |
| UNQ733/PRO1419/C4orf7/FDC-SP | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 260436 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction