Learn More
FBXW11, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FBXW11.
Brand: Abnova PAB10237.100ug
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbws class and, in addition to an F-box, contains multiple WD40 repeats. This gene contains at least 14 exons, and its alternative splicing generates 3 transcript variants diverging at the presence/absence of two alternate exons. [provided by RefSeq]
Specifications
| FBXW11 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of FBXW11. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| NP_387449;Q9UKB1 | |
| FBXW11 | |
| A synthetic peptide corresponding to N-terminus of human FBXW11. | |
| RUO | |
| 23291 | |
| Bovine, Canine, Chicken, Chimpanzee, Equine, Human, Mouse | |
| Liquid |
| ELISA, Immunofluorescence, Western Blot | |
| Unconjugated | |
| ELISA (1:10000-1:100000) Western Blot (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| FBXW11 | |
| BTRC2/BTRCP2/FBW1B/FBXW1B/Fbw11/Hos/KIAA0696 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody reacts with human betaTrCP2 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only