Learn More
FBXO11, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FBXO11.
Brand: Abnova PAB28680.100uL
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: GHRAKRARVSGKSQDLSAAPAEQYLQEKLPDEVVLKIFSYLLEQDLCRAACVCKRFSELANDPILWKRLYMEVFEYTRPMMHPEPGKFYQINPEEYEHPNPWKESFQQLYKGAHVKPGFAEHFYSNPARYKGRSpecifications
| FBXO11 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FBXO11. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| FBX11/FLJ12673/MGC44383/PRMT9/UBR6/UG063H01/VIT1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 80204 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| FBXO11 | |
| FBXO11 | |
| Recombinant protein corresponding to amino acids of human FBXO11. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only