missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBRS, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FBRS.
Brand: Abnova PAB24061.100uL
This item is not returnable.
View return policy
Description
Fibrosin is a lymphokine secreted by activated lymphocytes that induces fibroblast proliferation (Prakash and Robbins, 1998 [PubMed 9809749]).[supplied by OMIM
Sequence: LRHQEKMKGDSHKLDFRNDLLPCLPGPYGALPPGQELSHPASLFTATGAVHAAANPFTAAPGAHGPFLSPSTHSpecifications
| FBRS | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FBRS. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9HAH7 | |
| FBRS | |
| Recombinant protein corresponding to amino acids of human FBRS. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| FBRS | |
| FBS/FBS1/FLJ11618 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 64319 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction