Learn More
FBN2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FBN2.
Brand: Abnova PAB20628.100uL
Description
The protein encoded by this gene is a component of connective tissue microfibrils and may be involved in elastic fiber assembly. Mutations in this gene cause congenital contractural arachnodactyly. [provided by RefSeq
Sequence: GMCFSGLVNGRCAQELPGRMTKMQCCCEPGRCWGIGTIPEACPVRGSEEYRRLCMDGLPMGGIPGSAGSRPGGTGGNGFAPSGNGNGYGPGGTGFIPIPGGNGFSPGVGGAGVGAGGQGPIITGLTILNQSpecifications
| FBN2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FBN2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P35556 | |
| FBN2 | |
| Recombinant protein corresponding to amino acids of human FBN2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| FBN2 | |
| CCA/DA9 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2201 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only