missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fasl, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a synthetic peptide of Fasl.
Brand: Abnova PAB0844.200ug
This item is not returnable.
View return policy
Specifications
| Fasl | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a synthetic peptide of Fasl. | |
| In PBS (0.2% gelatin, 0.09% sodium azide) | |
| Apt1Lg1/Fasl/Tnfsf6 | |
| Rabbit | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (Frozen), Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Faslg | |
| Faslg | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of rat Fasl. | |
| RUO | |
| 25385 | |
| Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction