Learn More
FANCG Rabbit anti-Chimpanzee, Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FANCG.
Brand: Abnova PAB10048.100ug
Description
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group G. [provided by RefSeq
Specifications
| FANCG | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of FANCG. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| NP_004620;O15287 | |
| FANCG | |
| A synthetic peptide corresponding to amino acids 1-12 of human FANCG. | |
| RUO | |
| 2189 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:10000-1:50000) Western Blot (1:500-1:2000) Immunohistochemistry (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| FANCG | |
| FAG/XRCC9 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Chimpanzee, Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only