Learn More
FANCA Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FANCA.
Brand: Abnova PAB12429.100uL
Description
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group A. Alternative splicing results in multiple transcript variants encoding different isoforms. Mutations in this gene are the most common cause of Fanconi anemia. [provided by RefSeq
Specifications
| FANCA | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of FANCA. | |
| In antiserum | |
| O15360 | |
| FANCA | |
| A synthetic peptide corresponding to amino acids 1200-1300 of human FANCA. | |
| RUO | |
| 2175 | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| FANCA | |
| FA/FA-H/FA1/FAA/FACA/FAH/FANCH/MGC75158 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only