missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM48A Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FAM48A.
Brand: Abnova PAB23360.100uL
This item is not returnable.
View return policy
Description
Sequence: DSETIRLPYEEGELLEYLDAEELPPILVDLLEKSQVNIFHCGCVIAEIRDYRQSSNMKSPGYQSRHILLRPTMQTLICDVHSITSDNHKWTQEDSpecifications
| FAM48A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FAM48A. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q8NEM7 | |
| FAM48A | |
| Recombinant protein corresponding to amino acids of human FAM48A. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| FAM48A | |
| C13/C13orf19/FP757/P38IP/bA421P11.4 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 55578 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction