missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM129A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FAM129A.
Brand: Abnova PAB26827.100ug
This item is not returnable.
View return policy
Description
Sequence: KDDVKEGEGGQESSpecifications
| FAM129A | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| FAM129A | |
| C1orf24/FLJ38228/GIG39/NIBAN | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116496 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of FAM129A. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| Q9BZQ8 | |
| FAM129A | |
| A synthetic peptide corresponding to human FAM129A. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction