Learn More
FAF1 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FAF1.
Brand: Abnova PAB18292.100ug
Description
Interaction of Fas ligand (TNFSF6) with the FAS antigen (TNFRSF6) mediates programmed cell death, also called apoptosis, in a number of organ systems. The protein encoded by this gene binds to FAS antigen and can initiate apoptosis or enhance apoptosis initiated through FAS antigen. Initiation of apoptosis by the protein encoded by this gene requires a ubiquitin-like domain but not the FAS-binding domain. [provided by RefSeq
Specifications
| FAF1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| FAF1 | |
| FAF1 | |
| A synthetic peptide corresponding to human FAF1. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Lyophilized |
| Western Blot | |
| 0.5 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of FAF1. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| CGI-03/FLJ37524/HFAF1s/UBXD12/UBXN3A/hFAF1 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 11124 | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only