missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F13A1/F13A2/F13A3/F13B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native F13A1/F13A2/F13A3/F13B.
Brand: Abnova PAB19873.150ug
This item is not returnable.
View return policy
Specifications
| F13A1/F13A2/F13A3/F13B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native F13A1/F13A2/F13A3/F13B. | |
| In PBS, pH 7.5 (0.02% sodium azide, 50% glycerol (v/v)) | |
| F13A1 | |
| Native purified F13A1/F13A2/F13A3/F13B from human plasma. | |
| 150 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| F13A | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 2162/2163/2164/2165 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction