Learn More
EYA4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant EYA4.
Brand: Abnova PAB23343.100uL
Description
This gene encodes a member of the eyes absent (EYA) family of proteins. The encoded protein may act as a transcriptional activator through its protein phosphatase activity, and it may be important for eye development, and for continued function of the mature organ of Corti. Mutations in this gene are associated with postlingual, progressive, autosomal dominant hearing loss at the deafness, autosomal dominant nonsyndromic sensorineural 10 locus. Defects in this gene are also associated with dilated cardiomyopathy 1J. Three transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: TGGENMTVLNTADWLLSCNTPSSATMSLLAVKTEPLNSSETTATTGDGALDTFTGSVITSSGYSPRSASpecifications
| EYA4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant EYA4. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| O95677 | |
| EYA4 | |
| Recombinant protein corresponding to amino acids of human EYA4. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| EYA4 | |
| CMD1J/DFNA10 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2070 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only