Learn More
EXOC6, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant EXOC6.
Brand: Abnova PAB22976.100uL
Description
The product of this gene belongs to the SEC15 family. It is highly similar to the protein encoded by Saccharomyces cerevisiae SEC15 gene. This protein is essential for vesicular traffic from the Golgi apparatus to the cell surface in yeast. It is one of the components of a multiprotein complex required for exocytosis. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Sequence: TFSVSLQKQNKMKFGKNMYINRDRIPEERNETVLKHSLEEESpecifications
| EXOC6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant EXOC6. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q8TAG9 | |
| EXOC6 | |
| Recombinant protein corresponding to amino acids of human EXOC6. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| EXOC6 | |
| DKFZp761I2124/EXOC6A/FLJ1125/FLJ11251/MGC33397/SEC15/SEC15L/SEC15L1/SEC15L3/Sec15p | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 54536 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only