Learn More
ESR1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ESR1.
Brand: Abnova PAB27451.100ug
Description
This gene encodes an estrogen receptor, a ligand-activated transcription factor composed of several domains important for hormone binding, DNA binding, and activation of transcription. The protein localizes to the nucleus where it may form a homodimer or a heterodimer with estrogen receptor 2. Estrogen and its receptors are essential for sexual development and reproductive function, but also play a role in other tissues such as bone. Estrogen receptors are also involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis. Alternative splicing results in several transcript variants, which differ in their 5' UTRs and use different promoters. [provided by RefSeq]
Specifications
| ESR1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| ESR1 | |
| DKFZp686N23123/ER/ESR/ESRA/Era/NR3A1 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 2099 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of ESR1. | |
| In PBS (0.02% sodium azide) | |
| NP_001116214 | |
| ESR1 | |
| A synthetic peptide corresponding to 18 amino acids at internal region of human ESR1. | |
| 100 μg | |
| Primary | |
| ESR1 antibody is human, mouse and rat reactive. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only