Learn More
ERN1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ERN1.
Brand: Abnova PAB12433.100uL
Description
The protein encoded by this gene is the ER to nucleus signalling 1 protein, a human homologue of the yeast Ire1 gene product. This protein possesses intrinsic kinase activity and an endoribonuclease activity and it is important in altering gene expression as a response to endoplasmic reticulum-based stress signals. [provided by RefSeq
Specifications
| ERN1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ERN1. | |
| In PBS | |
| O75460 | |
| ERN1 | |
| A synthetic peptide corresponding to amino acids 700-800 of human ERN1. | |
| RUO | |
| 2081 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| ERN1 | |
| FLJ30999/IRE1/IRE1P/MGC163277/MGC163279 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only