Learn More
ERBB4 Rabbit anti-Hamster, Horse, Human, Monkey, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ERBB4.
Brand: Abnova PAB16286.50ug
Description
This gene is a member of the Tyr protein kinase family and the epidermal growth factor receptor subfamily. It encodes a single-pass type I membrane protein with multiple cysteine rich domains, a transmembrane domain, a tyrosine kinase domain, a phosphotidylinositol-3 kinase binding site and a PDZ domain binding motif. The protein binds to and is activated by neuregulins and other factors and induces a variety of cellular responses including mitogenesis and differentiation. Multiple proteolytic events allow for the release of a cytoplasmic fragment and an extracellular fragment. Mutations in this gene have been associated with cancer. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq
Specifications
| ERBB4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ERBB4. | |
| In PBS (0.09% sodium azide) | |
| Q15303 | |
| ERBB4 | |
| A synthetic peptide (conjugated with KLH) corresponding to human ERBB4. | |
| 50 μg | |
| Primary | |
| Internal domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15-30 ug/mL) The optimal working dilution should be determined by the end user. | |
| ERBB4 | |
| HER4/MGC138404/p180erbB4 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 2066 | |
| Equine, Hamster, Human, Monkey, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only