Learn More
ERBB2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ERBB2.
Brand: Abnova PAB28423.100uL
Description
This gene encodes a member of the epidermal growth factor (EGF) receptor family of receptor tyrosine kinases. This protein has no ligand binding domain of its own and therefore cannot bind growth factors. However, it does bind tightly to other ligand-bound EGF receptor family members to form a heterodimer, stabilizing ligand binding and enhancing kinase-mediated activation of downstream signalling pathways, such as those involving mitogen-activated protein kinase and phosphatidylinositol-3 kinase. Allelic variations at amino acid positions 654 and 655 of isoform a (positions 624 and 625 of isoform b) have been reported, with the most common allele, Ile654/Ile655, shown here. Amplification and/or overexpression of this gene has been reported in numerous cancers, including breast and ovarian tumors. Alternative splicing results in several additional transcript variants, some encoding different isoforms and others that have not been fully characterized. [provided by RefSeq
Sequence: LPSETDGYVAPLTCSPQPEYVNQPDVRPQPPSPREGPLPAARPAGATLERPKTLSPGKNGVVKDVFAFGGAVENPEYLTPQGGAAPQPHPPPAFSPAFDNLYYWDQDPPERGAPPSTFKGTPTASpecifications
| ERBB2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ERBB2. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| B4DTR1 | |
| ERBB2 | |
| Recombinant protein corresponding to human ERBB2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:100-1:250) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| ERBB2 | |
| CD340/HER-2/HER-2/neu/HER2/NEU/NGL/TKR1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2064 | |
| Store at 4 °C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only