Learn More
EPS15 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant EPS15.
Brand: Abnova PAB20429.100uL
Description
This gene encodes a protein that is part of the EGFR pathway. The protein is present at clatherin-coated pits and is involved in receptor-mediated endocytosis of EGF. Notably, this gene is rearranged with the HRX/ALL/MLL gene in acute myelogeneous leukemias. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq
Sequence: CFFRQSTDPFATSSTDPFSAANNSSITSVETLKHNDPFAPGGTVVAASDSATDPFASVFGNESFGGGFADFSTLSKVNNEDPFRSATSSSVSNVVITKNVFEETSVKSEDEPPALPPKIGTPTRPCPLPPGKRSINKLDSPDSpecifications
| EPS15 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant EPS15. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P42566 | |
| EPS15 | |
| Recombinant protein corresponding to amino acids of human EPS15. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| EPS15 | |
| AF-1P/AF1P/MLLT5 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2060 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only