Learn More
EPHA3 Rabbit anti-Hamster, Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EPHA3.
Brand: Abnova PAB3006.400uL
Description
This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene encodes a protein that binds ephrin-A ligands. Two alternatively spliced transcript variants have been described for this gene. [provided by RefSeq
Specifications
| EPHA3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of EPHA3. | |
| In PBS (0.09% sodium azide) | |
| P29320 | |
| EPHA3 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human EPHA3. | |
| 400 μL | |
| Primary | |
| Hamster, Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| EPHA3 | |
| ETK/ETK1/HEK/HEK4/TYRO4 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 2042 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only